The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013516.2 | NP_038544.1 | 3 - 4 | 389 | 149 |
GenBank mRNA | BC139323.1 | - | 3 - 4 | 313 | 149 |
BC139324.1 | - | 3 - 4 | 313 | 149 | |
J05019.1 | - | 3 - 4 | 313 | 149 |