The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013586.4 | NP_038614.2 | 13 - 14 | 2285 | 56 |
GenBank mRNA | AF053368.1 | - | 13 - 14 | 2316 | 56 |
AK030548.1 | - | 13 - 14 | 2286 | 56 | |
BC011298.1 | - | 13 - 14 | 2279 | 56 |