The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019731.1 | NP_062705.1 | 1 - 2 | 108 | 75 |
GenBank mRNA | AF153451.1 | - | 1 - 2 | 108 | 75 |
AF288690.1 | - | 1 - 2 | 121 | 75 | |
AK011573.1 | - | 1 - 2 | 113 | 75 | |
AK012805.1 | - | 1 - 2 | 144 | 75 | |
AK017724.1 | - | 1 - 2 | 124 | 75 | |
BC027277.1 | - | 1 - 2 | 126 | 75 |