The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164311.1 | NP_001157783.1 | 14 - 15 | 2313 | 63 |
NM_053083.3 | NP_444313.3 | 14 - 15 | 2310 | 63 | |
GenBank mRNA | AF338440.1 | - | 13 - 14 | 2205 | 63 |
AK014331.1 | - | 14 - 15 | 2312 | 63 | |
BC068307.1 | - | 14 - 15 | 2252 | 63 |