Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkcsh
Entrez Gene ID
19089
Gene Name
protein kinase C substrate 80K-H
Gene Aliases
80K-H, PKCSH
Chromosome Location
Chr.9: 22002988 - 22014245 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293650.1 NP_001280579.1 7 - 8 761 71
NM_001293651.1 NP_001280580.1 7 - 8 765 71
NM_008925.2 NP_032951.1 7 - 8 761 71
GenBank mRNA AK137661.1 - 7 - 8 728 71
AK153931.1 - 7 - 8 736 71
AK159705.1 - 7 - 8 732 71
BC009816.1 - 7 - 8 694 71
U92794.1 - 7 - 8 714 71

View Full Product Details