The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011706.2 | NP_035836.2 | 3 - 4 | 563 | 95 |
GenBank mRNA | AB021665.1 | - | 3 - 4 | 645 | 95 |
AK008689.1 | - | 1 - 2 | 88 | 95 | |
AK089004.1 | - | 4 - 5 | 578 | 95 | |
AK134052.1 | - | 2 - 3 | 876 | 95 | |
AK162841.1 | - | 2 - 3 | 437 | 95 | |
AK171222.1 | - | 2 - 3 | 486 | 95 | |
AK200065.1 | - | 1 - 2 | 151 | 95 | |
AK217648.1 | - | 1 - 2 | 62 | 95 | |
BB631765.1 | - | 3 - 4 | 563 | 95 | |
BC005415.1 | - | 2 - 3 | 510 | 95 |