The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290707.1 | NP_001277636.1 | 19 - 20 | 2362 | 70 |
NM_011866.2 | NP_035996.2 | 18 - 19 | 2316 | 70 | |
GenBank mRNA | AF110507.1 | - | 18 - 19 | 2187 | 70 |
AK039249.1 | - | 19 - 20 | 2362 | 70 | |
AK162221.1 | - | 18 - 19 | 1971 | 70 | |
AK162804.1 | - | 16 - 17 | 1857 | 70 | |
AK166310.1 | - | 10 - 11 | 1328 | 70 | |
AY360383.1 | - | 18 - 19 | 2316 | 70 | |
BC113201.1 | - | 18 - 19 | 2182 | 70 |