The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011890.4 | NP_036020.1 | 3 - 4 | 449 | 69 |
GenBank mRNA | AB024921.1 | - | 3 - 4 | 463 | 69 |
AK014381.2 | - | 3 - 4 | 463 | 69 | |
AK132306.1 | - | 3 - 4 | 508 | 69 | |
AK169403.1 | - | 3 - 4 | 450 | 69 | |
BC052349.1 | - | 3 - 4 | 439 | 69 |