The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019549.2 | NP_062422.1 | 6 - 7 | 882 | 140 |
GenBank mRNA | AF073294.1 | - | 6 - 7 | 866 | 140 |
AF181829.1 | - | 6 - 7 | 826 | 140 | |
AF303745.1 | - | 7 - 8 | 814 | 140 | |
AK033872.1 | - | 6 - 7 | 820 | 140 | |
AK038804.1 | - | - | 920 | 140 | |
AK050340.1 | - | 6 - 7 | 824 | 140 | |
AK075810.1 | - | 6 - 7 | 826 | 140 | |
AK138831.1 | - | 6 - 7 | 820 | 140 | |
AK151460.1 | - | 6 - 7 | 794 | 140 | |
AK152501.1 | - | 6 - 7 | 794 | 140 | |
AK153109.1 | - | 6 - 7 | 794 | 140 | |
AK156235.1 | - | 6 - 7 | 883 | 140 | |
BC066185.1 | - | 6 - 7 | 808 | 140 |