Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pkp3
Entrez Gene ID
56460
Gene Name
plakophilin 3
Gene Aliases
2310056L12Rik
Chromosome Location
Chr.7: 141078229 - 141090510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162924.1 NP_001156396.1 6 - 7 1440 63
NM_019762.2 NP_062736.2 5 - 6 1371 63
GenBank mRNA AF136719.1 - 5 - 6 1362 63
BC020472.1 - 5 - 6 1343 63
BC032045.1 - 5 - 6 1331 63
BC090668.1 - 6 - 7 1440 63
BC106141.1 - 5 - 6 1336 63
BC139775.1 - 5 - 6 1327 63

View Full Product Details