Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pkp3
Entrez Gene ID
56460
Gene Name
plakophilin 3
Gene Aliases
2310056L12Rik
Chromosome Location
Chr.7: 141078229 - 141090510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162924.1 NP_001156396.1 10 - 11 2094 145
NM_019762.2 NP_062736.2 9 - 10 2025 145
GenBank mRNA AF136719.1 - 9 - 10 2016 145
AK009943.1 - 3 - 4 386 145
AK154376.1 - 3 - 4 616 145
BC090668.1 - 10 - 11 2094 145
BC139775.1 - 9 - 10 1981 145

View Full Product Details