The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290180.1 | NP_001277109.1 | 2 - 3 | 285 | 95 |
NM_001290181.1 | NP_001277110.1 | 3 - 4 | 2179 | 95 | |
NM_024437.4 | NP_077757.2 | 3 - 4 | 439 | 95 | |
GenBank mRNA | AA616044.1 | - | 2 - 3 | 281 | 95 |
AF338424.1 | - | 3 - 4 | 399 | 95 | |
AK004924.1 | - | 3 - 4 | 2135 | 95 | |
AK088191.1 | - | 2 - 3 | 287 | 95 | |
BC069843.1 | - | 3 - 4 | 395 | 95 | |
CA460240.1 | - | 3 - 4 | 471 | 95 |