Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsta3
Entrez Gene ID
22122
Gene Name
tissue specific transplantation antigen P35B
Gene Aliases
AI256181, FX, Tstap35b
Chromosome Location
Chr.15: 75924669 - 75929821 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031201.1 NP_112478.1 2 - 3 235 68
GenBank mRNA AK029632.1 - 2 - 3 259 68
BC089003.1 - 1 - 2 171 68
M30127.2 - 2 - 3 235 68

View Full Product Details