Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgk3
Entrez Gene ID
170755
Gene Name
serum/glucocorticoid regulated kinase 3
Gene Aliases
2510015P22Rik, A330005P07Rik, Cisk, fy, fz
Chromosome Location
Chr.1: 9798130 - 9902570 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133220.2 NP_573483.1 2 - 3 228 114
GenBank mRNA AF312007.1 - 2 - 3 170 114
AK028722.1 - 2 - 3 173 114
AK031133.1 - 2 - 3 209 114
AK143836.1 - 2 - 3 216 114
AK151181.1 - 2 - 3 173 114
AK151867.1 - 2 - 3 205 114
AK151996.1 - 2 - 3 154 114

View Full Product Details