Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myt1
Entrez Gene ID
17932
Gene Name
myelin transcription factor 1
Gene Aliases
Nzf2, Nzf2a, Nzf2b, Nztf2
Chromosome Location
Chr.2: 181763332 - 181827781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171615.1 NP_001165086.1 17 - 18 2788 81
NM_001171616.1 NP_001165087.1 16 - 17 2898 81
NM_001171680.1 NP_001165151.1 11 - 12 2978 81
NM_008665.4 NP_032691.2 17 - 18 3012 81
GenBank mRNA AB082378.1 - 17 - 18 2999 81
AB093267.2 - 16 - 17 2605 81
AK133358.1 - 9 - 10 1577 81
AK135131.1 - 9 - 10 1174 81
BC063252.1 - 17 - 18 2788 81

View Full Product Details