The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010930.4 | NP_035060.1 | 2 - 3 | 337 | 61 |
GenBank mRNA | AK039481.1 | - | 2 - 3 | 328 | 61 |
AK081310.1 | - | 2 - 3 | 356 | 61 | |
AK081944.1 | - | 2 - 3 | 563 | 61 | |
AK136407.1 | - | 2 - 3 | 356 | 61 | |
AK155048.1 | - | 2 - 3 | 356 | 61 | |
BC003774.1 | - | 2 - 3 | 343 | 61 | |
X96585.1 | - | 2 - 3 | 337 | 61 | |
Y09257.1 | - | 2 - 3 | 325 | 61 |