The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013718.2 | NP_038746.1 | 4 - 5 | 519 | 105 |
GenBank mRNA | AF041433.1 | - | 4 - 5 | 510 | 105 |
AK151747.1 | - | 4 - 5 | 531 | 105 | |
AK168518.1 | - | 4 - 5 | 104 | ||
BC003736.1 | - | 4 - 5 | 519 | 105 |