Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss30
Entrez Gene ID
30943
Gene Name
protease, serine 30
Gene Aliases
2010015P21Rik, Disp, Disp1, Tmprss8
Chromosome Location
Chr.17: 23972127 - 23980572 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013921.3 NP_038949.2 5 - 6 727 110
GenBank mRNA BC010970.1 - 5 - 6 720 110
BC040348.1 - 5 - 6 727 110

View Full Product Details