Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc41
Entrez Gene ID
103220
Gene Name
tetratricopeptide repeat domain 41
Gene Aliases
AI449705, AU041509, Gnn
Chromosome Location
Chr.10: 86705632 - 86776844 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003939.2 NP_001003939.2 1 - 2 205 107
GenBank mRNA BC030307.1 - 1 - 2 183 107

View Full Product Details