Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abhd12
Entrez Gene ID
76192
Gene Name
abhydrolase domain containing 12
Gene Aliases
1500011G07Rik, 6330583M11Rik, AI431047, AW547313
Chromosome Location
Chr.2: 150832493 - 150904745 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC002263.1 - 1 - 2 60 83

View Full Product Details