The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025770.3 | NP_080046.3 | 5 - 6 | 498 | 87 |
GenBank mRNA | AB079383.1 | - | 4 - 5 | 454 | 87 |
AB091691.1 | - | 4 - 5 | 475 | 87 | |
AK009076.1 | - | 5 - 6 | 482 | 87 | |
AK017355.1 | - | 5 - 6 | 480 | 87 | |
AK077333.1 | - | 5 - 6 | 471 | 87 | |
BC023455.1 | - | 4 - 5 | 474 | 87 |