Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tctn2
Entrez Gene ID
67978
Gene Name
tectonic family member 2
Gene Aliases
4432405B04Rik, Tect2
Chromosome Location
Chr.5: 124598749 - 124627738 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026486.3 NP_080762.1 16 - 17 2013 69
GenBank mRNA AK012720.1 - 16 - 17 1989 69
AK014476.1 - 16 - 17 1979 69
AK031829.1 - 16 - 17 2014 69
AK032378.1 - 15 - 16 2098 69
BC058375.1 - 16 - 17 1876 69
DQ278869.1 - 16 - 17 2011 69

View Full Product Details