The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026487.3 | NP_080763.2 | 6 - 7 | 732 | 59 |
GenBank mRNA | AK007147.1 | - | 2 - 3 | 189 | 59 |
AK009419.1 | - | 2 - 3 | 320 | 59 | |
AK014967.1 | - | 6 - 7 | 749 | 59 | |
AK030719.1 | - | 6 - 7 | 752 | 59 | |
AK033639.1 | - | 6 - 7 | 756 | 59 | |
AK044925.1 | - | 6 - 7 | 732 | 59 | |
AK150469.1 | - | 6 - 7 | 752 | 59 | |
AK152059.1 | - | 6 - 7 | 752 | 59 | |
AK165953.1 | - | 6 - 7 | 767 | 59 | |
BC029085.1 | - | 6 - 7 | 732 | 59 | |
BC043051.1 | - | 6 - 7 | 717 | 59 |