The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027790.2 | NP_082066.2 | 7 - 8 | 723 | 61 |
GenBank mRNA | AK017274.1 | - | 7 - 8 | 725 | 61 |
BC117731.2 | - | 6 - 7 | 616 | 61 |