The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028513.3 | NP_082789.1 | 1 - 1 | 1107 | 106 |
GenBank mRNA | AK006772.1 | - | 1 - 1 | 952 | 106 |
AK077010.2 | - | 1 - 1 | 1109 | 106 | |
BC052344.1 | - | 1 - 1 | 973 | 106 |