The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052994.2 | NP_443720.1 | 4 - 5 | 567 | 61 |
GenBank mRNA | AJ278999.1 | - | 4 - 5 | 569 | 61 |
AK122233.1 | - | 4 - 5 | 229 | 61 | |
AK140483.1 | - | 4 - 5 | 473 | 61 | |
AK162941.1 | - | 4 - 5 | 472 | 61 | |
BC057324.1 | - | 4 - 5 | 567 | 61 | |
BC060210.1 | - | 4 - 5 | 519 | 61 |