The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053128.2 | NP_444358.2 | 1 - 1 | 633 | 69 |
GenBank mRNA | AF326296.1 | - | 1 - 1 | 69 | |
AK030570.1 | - | 1 - 1 | 633 | 69 | |
AY013785.1 | - | 1 - 1 | 449 | 69 | |
BC079557.1 | - | 1 - 1 | 547 | 69 |