Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crip3
Entrez Gene ID
114570
Gene Name
cysteine-rich protein 3
Gene Aliases
TLP, TLP-A, TLP-B, Tlmp
Chromosome Location
Chr.17: 46428195 - 46431780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053250.2 NP_444480.1 5 - 6 412 70
NM_181664.2 NP_858050.1 5 - 6 412 70
GenBank mRNA AF367970.1 - 5 - 6 412 70
AF367971.1 - 5 - 6 412 70
AK080272.1 - 5 - 6 412 70
BC116360.1 - 5 - 6 412 70

View Full Product Details