Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Strc
Entrez Gene ID
140476
Gene Name
stereocilin
Gene Aliases
DFNB16
Chromosome Location
Chr.2: 121363248 - 121381370 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080459.2 NP_536707.2 23 - 24 4719 111
GenBank mRNA AF375593.1 - 23 - 24 4719 111

View Full Product Details