The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288578.1 | NP_001275507.1 | 8 - 9 | 1172 | 86 |
NM_007410.3 | NP_031436.2 | 8 - 9 | 1190 | 86 | |
GenBank mRNA | AK002790.1 | - | 8 - 9 | 1112 | 86 |
AK076507.1 | - | 8 - 9 | 1119 | 86 | |
AK146949.1 | - | 8 - 9 | 1134 | 86 | |
AK159803.1 | - | 8 - 9 | 1170 | 86 | |
BC062879.1 | - | 8 - 9 | 1116 | 86 | |
BC090978.1 | - | 8 - 9 | 1113 | 86 | |
M84147.1 | - | 8 - 9 | 1140 | 86 |