Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acrbp
Entrez Gene ID
54137
Gene Name
proacrosin binding protein
Gene Aliases
OY-TES-1, sp32
Chromosome Location
Chr.6: 125049927 - 125063265 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127340.1 NP_001120812.1 4 - 5 556 102
NM_016845.2 NP_058541.2 4 - 5 556 102
GenBank mRNA AK076691.1 - 4 - 5 521 102
AK132705.1 - 4 - 5 524 102
BC011079.1 - 4 - 5 794 102
BC114591.1 - 4 - 5 525 102
BM938496.1 - 3 - 4 329 102
D17573.1 - 4 - 5 508 102
D17574.1 - 4 - 5 560 102

View Full Product Details