Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acrbp
Entrez Gene ID
54137
Gene Name
proacrosin binding protein
Gene Aliases
OY-TES-1, sp32
Chromosome Location
Chr.6: 125049927 - 125063265 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016845.2 NP_058541.2 5 - 6 1019 68
GenBank mRNA AK132705.1 - 5 - 6 987 68
D17573.1 - 5 - 6 971 68

View Full Product Details