The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253381.1 | NP_001240310.1 | 5 - 6 | 1236 | 73 |
NM_017377.5 | NP_059073.1 | 5 - 6 | 1201 | 73 | |
NR_045570.1 | 6 - 7 | 1256 | 73 | ||
GenBank mRNA | AB019541.2 | - | 3 - 4 | 740 | 73 |
AF142670.1 | - | 4 - 5 | 845 | 73 | |
AK145250.1 | - | 6 - 7 | 1258 | 73 | |
AK165829.1 | - | 5 - 6 | 1195 | 73 | |
BC138567.1 | - | 4 - 5 | 837 | 73 | |
BC138569.1 | - | 4 - 5 | 837 | 73 | |
BC145335.1 | - | 4 - 5 | 872 | 73 |