The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039509.1 | NP_001034598.1 | 7 - 8 | 822 | 72 |
NM_019999.2 | NP_064383.1 | 6 - 7 | 731 | 72 | |
GenBank mRNA | AB041609.1 | - | 6 - 7 | 732 | 72 |
AK041238.1 | - | 6 - 7 | 665 | 72 | |
AK173125.1 | - | 7 - 8 | 796 | 72 | |
BC058945.1 | - | 8 - 9 | 918 | 72 |