The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022424.5 | NP_071869.2 | 5 - 6 | 904 | 93 |
GenBank mRNA | AB030187.1 | - | 1 - 2 | 76 | 93 |
AK013203.1 | - | 5 - 6 | 898 | 93 | |
BC139278.1 | - | 5 - 6 | 656 | 93 | |
BC139292.1 | - | 5 - 6 | 656 | 93 |