The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025611.5 | NP_079887.3 | 25 - 26 | 5138 | 70 |
GenBank mRNA | AK008655.1 | - | 2 - 3 | 326 | 70 |
AK010907.1 | - | 4 - 5 | 663 | 70 | |
AK147156.1 | - | 25 - 26 | 4896 | 70 | |
AK170808.1 | - | 25 - 26 | 4911 | 70 | |
AK172882.1 | - | 24 - 25 | 4211 | 70 | |
BC019645.1 | - | 15 - 16 | 2426 | 70 | |
BC024345.1 | - | 4 - 5 | 930 | 70 | |
BC026946.1 | - | 7 - 8 | 1246 | 70 | |
BC059865.1 | - | 21 - 22 | 3651 | 70 | |
BC065096.1 | - | 24 - 25 | 4625 | 70 | |
BC100544.1 | - | 4 - 5 | 774 | 70 | |
BC138998.1 | - | 25 - 26 | 5076 | 70 | |
BC139000.1 | - | 25 - 26 | 5076 | 70 |