The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025620.2 | NP_079896.1 | 1 - 1 | 450 | 82 |
GenBank mRNA | AK008892.1 | - | 1 - 1 | 451 | 82 |
AK008964.1 | - | 1 - 1 | 453 | 82 | |
BC024344.1 | - | 1 - 1 | 401 | 82 |