The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113564.1 | NP_001107036.1 | 6 - 7 | 1097 | 94 |
NM_001113565.1 | NP_001107037.1 | 6 - 7 | 1070 | 94 | |
NM_001113566.1 | NP_001107038.1 | 6 - 7 | 1052 | 94 | |
NM_025814.2 | NP_080090.2 | 6 - 7 | 1115 | 94 | |
GenBank mRNA | AK004678.1 | - | 6 - 7 | 1115 | 94 |
AK010860.1 | - | 6 - 7 | 1070 | 94 | |
AK011051.1 | - | 6 - 7 | 1084 | 94 | |
AK088278.1 | - | 6 - 7 | 1021 | 94 | |
AK135473.1 | - | 5 - 6 | 852 | 94 | |
AK144764.1 | - | 6 - 7 | 1095 | 94 | |
AK146418.1 | - | 6 - 7 | 1039 | 94 | |
AK149505.1 | - | 6 - 7 | 1024 | 94 | |
BC006030.1 | - | 6 - 7 | 1024 | 94 | |
BC013665.1 | - | 6 - 7 | 1006 | 94 | |
BC030502.1 | - | 6 - 7 | 1014 | 94 |