The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252627.1 | NP_001239556.1 | 7 - 8 | 1077 | 101 |
NM_001252628.1 | NP_001239557.1 | 8 - 9 | 1167 | 101 | |
NM_007663.3 | NP_031689.1 | 8 - 9 | 1167 | 101 | |
GenBank mRNA | AF016271.1 | - | 8 - 9 | 1035 | 101 |
AJ609635.1 | - | 8 - 9 | 1034 | 101 | |
AK052636.1 | - | 8 - 9 | 970 | 101 | |
AK164546.1 | - | 7 - 8 | 1078 | 101 | |
BC015251.1 | - | 8 - 9 | 1015 | 101 |