Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rb1
Entrez Gene ID
19645
Gene Name
retinoblastoma 1
Gene Aliases
Rb, Rb-1, pRb
Chromosome Location
Chr.14: 73194453 - 73325887 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009029.2 NP_033055.2 11 - 12 1255 102
GenBank mRNA AK011246.1 - 11 - 12 1255 102
AK141028.1 - 11 - 12 1287 102
AK146467.1 - 6 - 7 586 102
BC096525.1 - 11 - 12 1255 102
DQ400415.1 - 11 - 12 1105 102
M26391.1 - 11 - 12 1203 102

View Full Product Details