The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009837.1 | NP_033967.1 | 12 - 13 | 1650 | 85 |
GenBank mRNA | AK010634.1 | - | 12 - 13 | 1618 | 85 |
AK088703.1 | - | 2 - 3 | 295 | 85 | |
AK146242.1 | - | 12 - 13 | 1620 | 85 | |
AK150601.1 | - | 12 - 13 | 1597 | 85 | |
AK152173.1 | - | 12 - 13 | 1597 | 85 | |
AK152665.1 | - | 12 - 13 | 1618 | 85 | |
AK165229.1 | - | 12 - 13 | 1618 | 85 | |
AK167026.1 | - | 12 - 13 | 1618 | 85 | |
AK167262.1 | - | 12 - 13 | 1587 | 85 | |
AK169339.1 | - | 12 - 13 | 1597 | 85 | |
BC054773.1 | - | 12 - 13 | 1610 | 85 | |
L25913.1 | - | 12 - 13 | 1489 | 85 | |
Z31554.1 | - | 12 - 13 | 1650 | 85 |