The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013672.2 | NP_038700.2 | 2 - 3 | 265 | 75 |
GenBank mRNA | AF022363.1 | - | 2 - 3 | 253 | 75 |
AF062566.1 | - | 2 - 3 | 201 | 75 | |
CJ181598.1 | - | 2 - 3 | 272 | 75 |