The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001806.2 | NP_001001806.1 | 2 - 2 | 567 | 133 |
GenBank mRNA | AK142565.1 | - | - | 373 | 133 |
AK159513.1 | - | 2 - 2 | 568 | 133 | |
AK160955.1 | - | 2 - 2 | 566 | 133 | |
AK170763.1 | - | 2 - 2 | 568 | 133 | |
BC006189.1 | - | 1 - 1 | 416 | 133 | |
BC028330.1 | - | 2 - 2 | 525 | 133 | |
BC038863.1 | - | 2 - 2 | 289 | 133 | |
BC139416.1 | - | 2 - 2 | 443 | 133 | |
M58564.1 | - | 1 - 1 | 244 | 133 |