Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpst2
Entrez Gene ID
22022
Gene Name
protein-tyrosine sulfotransferase 2
Gene Aliases
AI448750, D5Ucla3, Tango13b, grm, grt
Chromosome Location
Chr.5: 112276682 - 112315356 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009419.3 NP_033445.2 4 - 5 1247 62
GenBank mRNA AF049890.1 - 3 - 4 1193 62
AK036436.1 - 3 - 4 1176 62
AK075672.1 - 4 - 5 1247 62
AK149502.1 - 4 - 5 1212 62
AK163461.1 - 4 - 5 1211 62
BC003721.1 - 4 - 5 1238 62

View Full Product Details