The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010325.2 | NP_034455.1 | 4 - 5 | 494 | 75 |
GenBank mRNA | AK098166.1 | - | 4 - 5 | 518 | 77 |
AK136556.1 | - | 4 - 5 | 531 | 75 | |
AK147953.1 | - | 4 - 5 | 534 | 75 | |
AK149886.1 | - | 4 - 5 | 531 | 75 | |
AK149926.1 | - | 4 - 5 | 531 | 75 | |
AK150194.1 | - | 3 - 4 | 250 | 75 | |
AK152921.1 | - | 4 - 5 | 531 | 75 | |
AK155075.1 | - | 4 - 5 | 525 | 75 | |
BC089015.1 | - | 4 - 5 | 520 | 75 | |
BC089341.1 | - | 4 - 5 | 532 | 75 | |
J02622.1 | - | 4 - 5 | 494 | 75 |