The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011657.2 | NP_035787.1 | 2 - 3 | 172 | 65 |
GenBank mRNA | AB016963.1 | - | 2 - 3 | 115 | 65 |
AF045582.1 | - | 2 - 3 | 125 | 65 | |
AK035363.1 | - | 2 - 3 | 137 | 65 | |
AK142527.1 | - | 2 - 3 | 162 | 65 | |
BC060068.1 | - | 2 - 3 | 102 | 65 | |
BY347356.1 | - | 2 - 3 | 175 | 65 |