Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp68
Entrez Gene ID
24135
Gene Name
zinc finger protein 68
Gene Aliases
KRAB3, KRAZ2, Zfp70, mszf22, mszf49
Chromosome Location
Chr.5: 138603652 - 138619743 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163797.1 NP_001157269.1 3 - 4 343 123
NM_013844.2 NP_038872.1 3 - 4 349 123
GenBank mRNA AB024005.1 - 3 - 4 333 123
AK013868.1 - 3 - 4 344 123
AK146092.1 - 3 - 4 310 123
AK153887.1 - 3 - 4 285 123

View Full Product Details