Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tgm1
Entrez Gene ID
21816
Gene Name
transglutaminase 1, K polypeptide
Gene Aliases
2310004J08Rik, Tgase1
Chromosome Location
Chr.14: 55700009 - 55713921 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161714.1 NP_001155186.1 5 - 6 989 74
NM_001161715.1 NP_001155187.1 5 - 6 1088 74
NM_019984.3 NP_064368.3 5 - 6 1037 74
GenBank mRNA AF186373.1 - 5 - 6 980 74
BC026422.1 - 5 - 6 1037 74

View Full Product Details