The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193303.1 | NP_001180232.1 | 7 - 8 | 966 | 65 |
NM_026148.3 | NP_080424.2 | 7 - 8 | 960 | 65 | |
NM_201242.1 | NP_957694.1 | 7 - 8 | 984 | 65 | |
GenBank mRNA | AK009390.1 | - | 7 - 8 | 967 | 65 |
AK014954.1 | - | 7 - 8 | 961 | 65 | |
AK082932.1 | - | 7 - 8 | 829 | 65 | |
AK088939.1 | - | 7 - 8 | 835 | 65 | |
AK151235.1 | - | 7 - 8 | 835 | 65 | |
AK151938.1 | - | 7 - 8 | 827 | 65 | |
AK152679.1 | - | 7 - 8 | 827 | 65 | |
AK153346.1 | - | 7 - 8 | 835 | 65 | |
BC005621.1 | - | 7 - 8 | 815 | 65 | |
BC067393.1 | - | 7 - 8 | 758 | 65 | |
DQ003608.1 | - | 7 - 8 | 951 | 65 | |
EU122953.1 | - | 7 - 8 | 950 | 65 | |
EU122954.1 | - | 7 - 8 | 818 | 65 |