The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026148.3 | NP_080424.2 | 9 - 10 | 1091 | 66 |
NM_201242.1 | NP_957694.1 | 9 - 10 | 1115 | 66 | |
GenBank mRNA | AK014954.1 | - | 9 - 10 | 1092 | 66 |
AK082932.1 | - | 9 - 10 | 960 | 66 | |
AK088939.1 | - | 9 - 10 | 966 | 66 | |
AK151235.1 | - | 9 - 10 | 966 | 66 | |
AK151938.1 | - | 9 - 10 | 958 | 66 | |
AK152679.1 | - | 9 - 10 | 958 | 66 | |
AK153346.1 | - | 9 - 10 | 966 | 66 | |
BC005621.1 | - | 9 - 10 | 946 | 66 | |
BC067393.1 | - | 9 - 10 | 889 | 66 | |
DQ003608.1 | - | 9 - 10 | 1082 | 66 | |
EU122953.1 | - | 9 - 10 | 1081 | 66 | |
EU122954.1 | - | 9 - 10 | 949 | 66 |